Master Lock® Adjustable Cable Lockout, NMC
# de catalogue 94007-602
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Description:.64cm (.25") steel cable with vinyl coating
- Cat. no.:94007-602
- Supplier No.:CABLO
Spécifications
A propos de cet article
Infinitely adjustable for a secure fit every time
Feed the cable end through the points to be locked out, then back through the lockout body. Tough, multi-strand .64cm (¼") dia. Steel cable with clear vinyl coating. Up to 4 workers can apply their locks. Includes a 15.24cm (6") cable.
Customers Who Bought This Also Bought