Adapter, [ST], Straight Hose Connection, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter features a straight hose connection with inner standard taper joint at bottom and glass stopcock.
- Straight hose connection
- Bottom inner [ST] joint
- Glass stopcock
- Fabricated from borosilicate glass
Adapter features a glass stopcock with a 2mm plug bore. Bore size represents the plug bore size.
Customers Who Bought This Also Bought