Adapter, [ST], Angled Hose Connection, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter features a 90 degree angled hose connection with inner standard taper joint at bottom and glass stopcock.
- 90° angled hose connection
- Bottom inner [ST] joint
- Glass stopcock
- Fabricated from borosilicate glass
Adapter features a glass stopcock with a 2mm or 3mm bore.
Customers Who Bought This Also Bought