Adapter. Distilling, Ace-Thred, Ace Glass Incorporated
# de catalogue 89051-308
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Distilling Ace-Thred Adapter
- Cat. no.:89051-308
- Supplier No.:5086-54
Spécifications
A propos de cet article
This adapter is designed for connecting distilling columns with a vertical condenser.
- Top Ace-Thred and thermometer
- Bottom [ST] inner joints
- Side arm at 75° angle
- Nylon bushing and FETFE o-ring included
- Fabricated from borosilicate glass
The top of this adapter has a #7 Ace-Thred and an adjustable length thermometer. The side arm is at 75 degrees, and the vertical side arm is 17.8cm from the center tube. Bottom joints are standard taper 24/40 inner. Supplied complete with 89051-122 nylon bushing and FETFE o-ring. For extra o-rings order 89060-958.
Customers Who Bought This Also Bought