Adapter, Offset, Ace Glass Incorporated
# de catalogue 89051-322
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Offset #7 Ace-Thred Adapter
- Cat. no.:89051-322
- Supplier No.:5101-54
Spécifications
A propos de cet article
This adapter features an Ace-Thred at the top, a reinforced standard taper outer joint on the side arm, and a standard taper inner joint at the bottom.
- Top Ace-Thred
- Side reinforced [ST] outer joint
- Bottom [ST] inner joint
- Nylon bushing and o-ring included
- Fabricated from borosilicate glass
Adapter comes with a #7 Ace-Thred at the top, reinforced standard taper 24/40 outer joint on side arm, and standard taper inner joint at bottom. Supplied with a 89051-122 nylon bushing and a 89060-958 FETFE O-Ring.
Customers Who Bought This Also Bought