Hose Connection Adapter, [ST], Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These adapters feature a straight hose connection at top.
- Bottom ground [ST] inner joint
- Top hose connection
- Fabricated from borosilicate glass
Adapters have a ground standard taper inner joint at the bottom and a straight hose connection at the top.
Customers Who Bought This Also Bought