Adapter, Septum Inlet, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This is a sampling adapter with a septum port at the top for handling air-sensitive materials.
- Septum port at top
- Bottom [ST] inner joint
- Septum included
- Fabricated from borosilicate glass
Featuring a standard taper inner joint at the bottom and septum port at the top. Supplied with a 89064-960 septum.
Customers Who Bought This Also Bought