Adapter, Kjeldahl Trap, Ace Glass Incorporated
# de catalogue 89051-448
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Kjeldahl Trap Adapter
- Cat. no.:89051-448
- Supplier No.:5226-10
Spécifications
A propos de cet article
The Kjeldahl trap adapter is used to prevent carryover of reactions that have a tendency to foam.
- Two ground [ST] inner joints
- Fabricated from borosilicate glass
This Kjeldahl Trap adapter has two 24/40 ground standard taper inner joints. The distance between the center of each joint is approximately 200mm.
Customers Who Bought This Also Bought