ContactUsLinkComponent
Adapter, Liquid Inlet, Ace Glass Incorporated
Adapter, Liquid Inlet, Ace Glass Incorporated
# de catalogue 89051-358
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
5175 ADAPTER, Liquid Inlet
Adapter, Liquid Inlet, Ace Glass Incorporated
# de catalogue 89051-358
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Liquid Inlet Adapter
  • Cat. no.:
    89051-358
  • Supplier No.:
    5175-10
Spécifications
A propos de cet article
This adapter has a standard taper inner joint and two vacuum take-offs.
  • [ST] inner joint
  • Two vacuum take-offs
  • Fabricated from borosilicate glass
Featuring a standard taper 24/40 inner joint. Use 3/8-inch I.D. tubing, size D hose connection.
Customers Who Bought This Also Bought