Adapter, Liquid Inlet, Ace Glass Incorporated
# de catalogue 89051-358
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Liquid Inlet Adapter
- Cat. no.:89051-358
- Supplier No.:5175-10
Spécifications
A propos de cet article
This adapter has a standard taper inner joint and two vacuum take-offs.
- [ST] inner joint
- Two vacuum take-offs
- Fabricated from borosilicate glass
Featuring a standard taper 24/40 inner joint. Use 3/8-inch I.D. tubing, size D hose connection.
Customers Who Bought This Also Bought