Adapter, Straight Tube, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter has a spherical socket joint at one end and a straight tube at the other end.
- Spherical socket joint
- Straight tube
- Fabricated from borosilicate glass
The socket joint to tubing adapter is sized for compression fittings.
Customers Who Bought This Also Bought