Adapter, PTFE, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter is used to connect a vial or a bottle to the vapor tube of a rotary evaporator.
- One end is [ST] outer joint with rib
- Female vial or bottle GPI thread on other end
- Fabricated from PTFE
One end of this adapter is a standard taper outer joint with rib for using clamp, while the other end is a female vial or bottle GPI thread. Joint connects to Thread. Thread size represents the GPI Thread Size.
Customers Who Bought This Also Bought