Adapter, Claisen, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter is available with glass or 1:5 solid PTFE stopcock plug on lower side arm.
- Top [ST] outer joint on center tube
- Accommodates thermometer
- Take-off arm at 75° angle
- Fabricated from borosilicate glass
The top joint on the center tube is a standard taper outer 10/30 for a 76mm immersion thermometer. All other joints are standard taper. Take-off arm is at a 75° angle to the vertical arm. Plug is 2mm bore. Plug style is Glass (89051-344 and 89064-822) and PTFE (89051-346).
Customers Who Bought This Also Bought