Adapter, Distilling Trap, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Distilling trap adapters are placed above flask to prevent carryover of reactions that have a tendency to foam.
- Top [ST] outer joint
- Bottom [ST] inner joint
- Fabricated from borosilicate glass
Adapter features a standard taper outer joint at the top and a standard taper inner joint at the bottom.
Customers Who Bought This Also Bought