Adapter, Spherical Joint, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These spherical joint adapters can be supplied to fit flask capacities of up to 72L.
- Can be modified to meet your application needs
- Hose connection is size C, for 3/8-inch ID tubing
- Fabricated from borosilicate glass
These spherical joint adapters can be modified to meet your application needs. You must specify the length needed below the joint, or the capacity or VWR Order Code of the vessel into which this adapter is being placed. The bottom tube will be supplied with the proper curvature, if required, to fit the vessel. Can be supplied to fit flask capacities of up to 72L. Hose connection is size C, for 3/8-inch ID tubing. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought