Adapter, Connecting, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This connecting adapter comes with standard taper or spherical joints.
- Standard taper or spherical joints
- Available in various lengths
- Fabricated from borosilicate glass
Featuring standard taper or spherical joints. Length values respresent the distance between the two joints.
Customers Who Bought This Also Bought