Adapter, Connecting, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter is used with vacuum manifolds as port connections.
- O-ring joint on one end
- Outer [ST] joint on opposite end
- One o-ring included
- Fabricated from borosilicate glass
Featuring a #15 o-ring joint on one end and an outer standard taper joint on the opposite end. Supplied with one o-ring.
Customers Who Bought This Also Bought