Adapter, Moisture Trap, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This is a unique adapter used in place of a Dean-Stark moisture test receiver.
- Top [ST] outer joint
- Bottom [ST] inner joint
- Side arm [ST] inner joint
- Fabricated from borosilicate glass
Simply add a condenser (not included) to the top standard taper outer joint, any graduated funnel from 125mL to 2000mL (not included) to bottom standard taper inner joint, attach a sample flask (not included) to the standard taper inner side arm joint, and you have a moisture test receiver.
Customers Who Bought This Also Bought