Adapter, Claisen, Ace Glass Incorporated
# de catalogue 89051-340
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:#7 Ace-Thred Claisen Adapter
- Cat. no.:89051-340
- Supplier No.:5136-54
Spécifications
A propos de cet article
The top side arm of this Claisen adapter has a #7 Ace-Thred for use with a nylon bushing for an adjustable length thermometer.
- Top reinforced outer [ST] joint
- Side and bottom inner [ST] joints
- Nylon bushing and FETFE o-ring included
- Fabricated from borosilicate glass
Supplied complete with a 89051-122 nylon bushing and 89060-958 FETFE o-ring. Side and bottom joints are 24/40 inner standard taper, top joint is reinforced 24/40 outer standard taper.
Customers Who Bought This Also Bought