Adapter, 'U', [ST] 24/40, Ace Glass
# de catalogue 89051-260
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:"U" Adapter
- Cat. no.:89051-260
- Supplier No.:5060-10
Spécifications
A propos de cet article
This is a connecting adapter with reinforced standard taper 24/40 outer joints at both ends.
- U-shaped
- Reinforced [ST] outer joints at both ends
- Fabricated from borosilicate glass
Customers Who Bought This Also Bought