Adapter, Twin, Ace Glass Incorporated
# de catalogue 89051-182
Fournisseur: Ace Glass
Small Business Enterprise
Spécifications
- Description:Twin Adapter
- Cat. no.:89051-182
- Supplier No.:5034-11
Spécifications
A propos de cet article
This adapter comes with an inner standard taper joint at bottom, one #7 Ace-Thred, and one 10/18 standard taper outer joint, offset at the top.
- Threaded opening
- Top Ace-Thred and [ST] outer joints, offset
- Bottom inner [ST] joint
- Bushing and o-ring included
- Fabricated from borosilicate glass
Threaded opening will accept 6.5 to 7mm diameter thermometers, bleed tubes, etc. The 24/25 medium length joint is compatible with a full length 24/40 joint. Supplied complete with 89051-122 nylon bushing and 89060-958 FETFE o-ring.
Customers Who Bought This Also Bought