Adapter, Straight Connecting, Borosilicate Glass, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This straight connecting adapter comes with either standard taper or spherical joints at both the top and the bottom.
- Inner joint on one end and outer joint on other
- Standard taper or spherical joints
- Fabricated from borosilicate glass
The length stated is the approximate overall length. Full-sized outer standard taper joints are reinforced.
Customers Who Bought This Also Bought