Adapter, Straight Connecting, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This straight connecting adapter has ground standard taper inner joints at both ends.
- Top ground [ST] inner joint
- Bottom ground [ST] inner joint
- Fabricated from borosilicate glass
Adapter is available in various lengths.
Customers Who Bought This Also Bought