Adapter, 'U', Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
This is a connecting adapter with spherical joints at both ends.
- U-shaped
- Spherical joints at both ends
- Fabricated from borosilicate glass
Length values respresent the distance between the two joints.
Customers Who Bought This Also Bought