Adapter, 105 Degree Side Arm, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter features a side arm at a 105° angle.
- Top and side reinforced [ST] outer joints
- Bottom [ST] inner joint
- With side arm
- Fabricated from borosilicate glass
This adapter comes with reinforced standard taper outer joints at the top and side, and an standard taper inner joint at the bottom. Side arm is at a 105 degree angle.
Customers Who Bought This Also Bought