Spatulas, Double End Mixing, Excelta Corp®
Fournisseur: Excelta
Certificats
A propos de cet article
These stainless steel spatulas are used for applying or scraping potting compound or wax.
- Double ended for flexibility
- Cleanroom safe, stainless steel
These large spatulas are primarily used for applying or scraping potting compound or wax. They can also be used for mixing epoxy.The 342 and 343 are made of stainless steel, 7" long.
Customers Who Bought This Also Bought