Adapter, 105 Degree Angle, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This 105° angle adapter has standard taper joints or spherical joints.
- 105° angle
- Top [ST] reinforced outer or socket joint
- Bottom [ST] inner or ball joint
- Fabricated from borosilicate glass
Adapter has standard taper joints (outer reinforced at top, inner at bottom) or spherical joints (socket at top, ball at bottom).
Customers Who Bought This Also Bought