Adapter, Distillate Take-Off, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This distillate take-off adapter has a 105° angle.
- Reinforced outer [ST] joint
- 105° angle
- Straight tube bottom
- Fabricated from borosilicate glass
Adapter comes with a reinforced outer standard taper joint, at a 105° angle. It has a straight tube bottom.
Customers Who Bought This Also Bought