Four Star™ Instrument Cleaning Brushes, EXCELTA®
Fournisseur: Excelta
Certificats
A propos de cet article
These instrument cleaning brushes are designed for general cleaning in tight areas.
- Handle is .25" in diameter
- Tip size of .13" wide and .19" long
The fibers are mechanically clamped into the stainless steel tube and inserted in the acetal handle. These brushes are used for scratch-free dry brushing.
Customers Who Bought This Also Bought