"Adapter, "Mini" #7 Ace-Thred, Ace Glass Incorporated"
Fournisseur: Ace Glass
Certificats
A propos de cet article
This "Mini" adapter has a ground joint at the bottom and a threaded nylon bushing with o-ring at the top that tightens into a #7 Ace-Thred joint to form a compression seal.
- Bottom ground joint
- Threaded nylon bushing with o-ring at top
- Accommodates thermometer
- Suitable for vacuum work
- Manufactured from borosilicate glass
Adapter features a threaded nylon bushing (complete with o-ring) at top that tightens into a #7 Ace-Thred joint to form a compression seal with thermometers, bleed tubes, etc. Standard taper 10/10 size will accommodate thermometers up to 6.4mm diameters; all others will accommodate 7mm diameter. This adapter is suitable for vacuum work.
Customers Who Bought This Also Bought