Adapter, Ace-Thred "Twin", Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter has an inner standard taper joint at bottom and two offset Ace-Thred ports at top.
- Bottom inner [ST] joint
- Two offset Ace-Thred ports at top
- Accommodates two inner tubes
- Two nylon bushings and FETFE o-rings included
Adapter has two threaded openings that enable you to insert two inner tubes, such as a thermometer and a bleed tube, through the same joint. Supplied complete with two nylon bushings and FETFE o-rings.
Customers Who Bought This Also Bought