Adapter, Straight Connecting with Glass Stopcock, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter comes with a glass plug stopcock and o-ring top joint, and either a standard taper or spherical joint at the bottom.
- Glass plug stopcock
- O-ring top joint
- Either [ST] or spherical bottom joint
- Fabricated from borosilicate glass
Adapter features a 4mm glass plug stopcock and #15 o-ring top joint.
Top o-ring is made of FETFE. FETFE is an exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along with excellent temperature and chemical compatibility. USP Class VI material.
Ordering information: Comes complete with 89061-002 FETFE o-ring for top joint.
Customers Who Bought This Also Bought