Adapter, PTFE, Ace-Thred, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This PTFE adapter is used to connect Ace-Thred to an inner standard taper joint.
- Fabricated from borosilicate glass
- One o-ring included
This adapter is supplied with one FETFE o-ring.
Customers Who Bought This Also Bought