Bushing, Nylon or PTFE, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This is a machine threaded nylon or PTFE bushing
Available with a hole in center or solid. For use with apparatus with a #7 internal threads.
Ordering information: Solid bushing is for drilling different size hole, only. NOT intended for use as a stopper. Nylon version has a maximum temperature of 100°C. PTFE version has a maximum temperature of 160°C.
Customers Who Bought This Also Bought