Tweezer, Bent General Purpose, Serrated, Excelta Corp®
Supplier: Avantor
Certificates
About this item
These are great tools for general assembly (large parts) and feature serrated tips and handles for positive gripping.
- Serrated grips for better control
- Serrated tips for gripping power
- Cleanroom safe, stainless steel construction
These 45° bent strong serrated tipped tweezer are ideal for general applications. They are made of anti-magnetic stainless steel and have serrated tips and grips for better control.
Frequently Bought Together