Adapter, Straight Connecting with PTFE Stopcock, Borosilicate Glass, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This straight connecting adapter has a PTFE stopcock plug.
- PTFE stopcock plug
- Available with [ST] inner and reinforced outer joints or spherical ball and socket joints
- Fabricated from borosilicate glass
Adapter features a 1:5 PTFE stopcock plug and standard taper inner and reinforced outer joints, or spherical ball and sockets joint, at top and bottom.
Customers Who Bought This Also Bought