Adapter, Straight Connecting with Glass Stopcock, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This straight connecting adapter is available with either a 2mm or 4mm bore glass stopcock plug.
- 2mm or 4mm bore glass stopcock plug
- Available with [ST] inner and reinforced outer joints or spherical ball and socket joints
- Fabricated from borosilicate glass
Available with standard taper inner and reinforced outer joints, or spherical ball and socket joints, at top and bottom.
Customers Who Bought This Also Bought