Tweezer, Ceramic Straight Fine Tip, Excelta Corp®
Supplier: Excelta
Certificates
About this item
Ceramic tweezers offer both heat resistant (2000 °F) for high temp applications and rigidity.
- Available with ergonomic grips
- Extremely hard tips
The 'Style 277-CE' white Zirconia ceramic tipped tweezer is 5.5" long. The tip size is 0.04" wide and the shank is made of nickel silver, which is non-magnetic. Zirconia ceramic is six times harder than stainless steel, non-magnetic and chemically inert. Solder will not adhere to the surface. They have a working temperature of over 2000 °F. The 'ET' suffix additionally features Ergo-Tweeze® soft cushioned ergonomic grips.
Customers Who Bought This Also Bought