MasterLock® Dual Jaw Aluminum Hasps, NMC
# de catalogue 94021-306
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Description:17.78 x 7.3 cm (7 x 2⅞")
- Cat. no.:94021-306
- Supplier No.:SL5
Spécifications
A propos de cet article
Made from aluminum, these hasps feature 1" and 1½" jaws, are spark resistant and hold up to 8 padlocks.
Customers Who Bought This Also Bought