Personal Protection Numbers and Symbols, National Marker
# de catalogue 94015-946
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Description:Personal protection numbers and symbols
- Material:PS Vinyl
- Cat. no.:94015-946
- Dimensions:152.4×101.6 mm (6×4")
- Supplier No.:HM41
Spécifications
A propos de cet article
An easy alternative to hand-writing information on labels.
Designed to be used with RTK labels. In the labels each number and symbols measures 0.5 and 0.75" respectively.
Customers Who Bought This Also Bought