MasterLock® Snap-On Advantage Hasp, NMC
# de catalogue 94021-302
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Cat. no.:94021-302
- Dimensions:5.08 x 14.92 cm (2 x 57/8")
- Supplier No.:SL115
Spécifications
A propos de cet article
Lock out your equipment and machinery in a snap
Features a spring loaded jaw end. Padlock inserted into the holes prevents the jaws from opening. Accommodates up to 5 padlocks. Hasp made from durable anodized aluminum and steel rivet construction.
Customers Who Bought This Also Bought