Drum Roller, Justrite®
# de catalogue 82011-470
Fournisseur: Justrite
Spécifications
- Description:Drum Roller
- Cat. no.:82011-470
- Supplier No.:25922
Spécifications
A propos de cet article
Facilitate movement of heavy, full 110 L (30 gal.) or 200 L (55 gal.) drums in vertical drum storage cabinets.
Customers Who Bought This Also Bought