Smokers Cease-Fire® Cigarette Butt Receptacle, Justrite®
Catalogue # 56610-466
Fournisseur: Justrite
Spécifications
- Dimensions:41.9dia. x 97.8H cm (141/2 x 381/2")
- Cat. no.:56610-466
- Supplier No.:26800
Spécifications
À propos de cet article
Flame-retardant, sturdy polyethylene receptacle with liner pail.
- Will not rust, dent, or crack
- Rain bonnet opening keeps rain out
- Lid lifts off to empty steel pail inside
Unique rain bonnet opening keeps rain out and discourages other trash. Simply lift off lid to empty steel pail inside. Tie-down notches offer security and greater stability under severe weather conditions.
Color: pewter gray