NMC Numbered Brass Valve Tags
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Great for jobs requiring numbering only
All tags are 0.035 Brass with black filled 1.27cm (1/2") numbers. 3.81cm (11/2") round tags.
Customers Who Bought This Also Bought