Master Lock® Rotating Gate Valve Lockouts, NMC
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Surrounds the valve operating handle to protect against accidental valve opening
Ideal for confined space applications. Made from tough, lightweight dielectric Xenoy® bodies.
Customers Who Bought This Also Bought