Target Organ Label, National Marker
# de catalogue 94015-950
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Description:Warning labels
- Material:PS Vinyl
- Cat. no.:94015-950
- Legend:Target organ
- Dimensions:152.4×63.5 mm (6×2.5")
- Supplier No.:HM46
Spécifications
A propos de cet article
It identify target organs and the route of entry of a hazardous substance.
Designed to be used with RTK labels.
Customers Who Bought This Also Bought