Technik™ Tapered Head Stainless Steel Cutters, Aven
Fournisseur: Aven Tools
Certificats
A propos de cet article
Cutters feature box joint construction and dual leaf springs for comfortable action.
- Made of mirror polished stainless steel
- ESD-safe grips
Cutting edges are induction hardened.
Customers Who Bought This Also Bought