SP Bel-Art Earth-Friendly '1001' Sampling Sticks, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
These earth-friendly sampling sticks are durably constructed with 100% PLA (polylactic acid) resin.
- PLA resin construction
- Earth-friendly with no petroleum-based materials
- Equally as strong as non-biodegradable plastic resins
PLA resin is a plant-based material and is a renewable resource with little environmental impact. PLA offers a lower carbon footprint, and is 100% biodegradable through industrial composting. Sampling sticks made from PLA polymers are just as strong as sampling sticks made from other non-biodegradable plastic resins. These earth-friendly sticks contain no petroleum-based materials and contribute far less greenhouse gases.
14.6L x 1.9W x 0.15H cm (5¾ x ¾ x ¹⁄₁₆")
Spécifications
- Catalog No:
- 89218-782
- Environmentally Preferable:
- Waste Reducing