Nalgene® Closures with Barbed Bulkhead Fittings, White PP, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Thermo Scientific Nalgene Closures with Barbed Bulkhead Fittings support the integration of tubing sets to Nalgene culture vessels.
Fittings may also be used with any Nalgene container with a 38-430 sized closure. The 38-430 closures are pre-drilled to accept either a 1/4 or 1/2 inch (6-13mm) ID fitting.
Customers Who Bought This Also Bought