Finnpipette® F-Stands, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
The Finnpipette® F-stand accommodates six pipettors
The sturdy crossbar structure with a slot for each pipettor ensures that pipetors will sit snugly in position. The Finnpipette® F-stand is suitable for both single channel and multichannel pipettors, and both F1 and F2 models. The single position multichannel stand is a multi-position stand that can accommodates any multichannel Finnpipette® pipettor. It allows storage in an upright or horizontal position.
Customers Who Bought This Also Bought