Finnpipette® Bottle Top Dispenser, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Finnpipette Dispensers protect users even when handling aggressive liquids
- Ergonomically designed
- Non-slip piston handle
- Excellent chemical and thermal resistance
- Precise self-locking system in the volume setting
The dispensers are available in six models, covering volumes between 0.2 and 60mL. There is a universal fit for reagent bottles, and the dispensing head cap prevents dripping.
Customers Who Bought This Also Bought